Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAGTTGCTTCTGTAACTCTCC[A/C]TTGAAACTGACTTCTCCCAAGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDR78 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR78 - WD repeat domain 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024763.4 | Intron | NP_079039.4 | ||||
NM_207014.2 | Intron | NP_996897.2 | ||||
XM_005271212.3 | Intron | XP_005271269.1 | ||||
XM_011542161.2 | Intron | XP_011540463.1 | ||||
XM_011542162.2 | Intron | XP_011540464.1 | ||||
XM_017002353.1 | Intron | XP_016857842.1 | ||||
XM_017002354.1 | Intron | XP_016857843.1 | ||||
XM_017002355.1 | Intron | XP_016857844.1 | ||||
XM_017002356.1 | Intron | XP_016857845.1 | ||||
XM_017002357.1 | Intron | XP_016857846.1 |