Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGAAAAAGGTGAACTGACTCAA[C/G]TAAAGTCTTCAGATTCGAACCCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf210 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf210 - chromosome 1 open reading frame 210 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164829.1 | Intron | NP_001158301.1 | ||||
NM_182517.2 | Intron | NP_872323.1 | ||||
XM_011540802.1 | Intron | XP_011539104.1 |
TMEM125 - transmembrane protein 125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |