Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCTTCTGCCCCAGGGTGAGGAC[A/G]AACAGAACTGAAGGGAGAGGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614259 MIM: 614443 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP57 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP57 - cilia and flagella associated protein 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EBNA1BP2 - EBNA1 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159936.1 | Intron | NP_001153408.1 | ||||
NM_006824.2 | Intron | NP_006815.2 |
FAM183A - family with sequence similarity 183 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6733 - microRNA 6733 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |