Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAGAGAAGGGAAAAGGGAAAC[C/T]TGTTGTTTAATGGGTGTGAAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM183A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM183A - family with sequence similarity 183 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101376.2 | 155 | Intron | NP_001094846.2 | |||
XM_005270875.4 | 155 | UTR 5 | XP_005270932.1 | |||
XM_005270876.4 | 155 | Intron | XP_005270933.1 | |||
XM_017001322.1 | 155 | Intron | XP_016856811.1 |