Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTGCTGGTGGGTCAACTGGATA[C/G]TCCTTTGGAAAGAAAAAATTAAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM183A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM183A - family with sequence similarity 183 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101376.2 | Intron | NP_001094846.2 | ||||
XM_005270875.4 | Intron | XP_005270932.1 | ||||
XM_005270876.4 | Intron | XP_005270933.1 | ||||
XM_017001322.1 | Intron | XP_016856811.1 |