Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCCAAAGTCCCCCTGAACCTCA[A/G]GTCGGAATCCAAAGCCCAGTAGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131241 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EDN2 - endothelin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302269.1 | Intron | NP_001289198.1 | ||||
NM_001956.4 | Intron | NP_001947.1 | ||||
XM_017000512.1 | Intron | XP_016856001.1 |
Set Membership: |
HapMap |