Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTGATCACTCTCACAGCATGT[A/G]TCCTATCCTTCGTCACATTCTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602135 MIM: 609365 MIM: 608241 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNALI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNALI1 - dynein axonemal light intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003462.3 | Intron | NP_003453.2 | ||||
XM_005271172.2 | Intron | XP_005271229.1 |
GNL2 - G protein nucleolar 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNIP1 - Smad nuclear interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |