Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGGAGACAAAAAACAAAGAGG[C/T]GGCTACTGTCACAGTACTTTATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607779 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AUNIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AUNIP - aurora kinase A and ninein interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAQR7 - progestin and adipoQ receptor family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178422.5 | 2845 | UTR 3 | NP_848509.1 | |||
XM_005245745.3 | 2845 | UTR 3 | XP_005245802.1 | |||
XM_005245746.3 | 2845 | UTR 3 | XP_005245803.1 | |||
XM_011540861.1 | 2845 | UTR 3 | XP_011539163.1 | |||
XM_011540862.2 | 2845 | UTR 3 | XP_011539164.1 |
Set Membership: |
HapMap |