Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTTTTTTGTTTGTTTTTGGTTT[G/T]GTTTTTGTTTTGAGACAAGGTCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604669 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKD2L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PKD2L2 - polycystin 2 like 2, transient receptor potential cation channel | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258448.1 | Intron | NP_001245377.1 | ||||
NM_001258449.1 | Intron | NP_001245378.1 | ||||
NM_001300921.1 | Intron | NP_001287850.1 | ||||
NM_014386.3 | Intron | NP_055201.2 | ||||
XM_011543318.2 | Intron | XP_011541620.1 | ||||
XM_011543321.1 | Intron | XP_011541623.1 | ||||
XM_017009343.1 | Intron | XP_016864832.1 | ||||
XM_017009344.1 | Intron | XP_016864833.1 | ||||
XM_017009345.1 | Intron | XP_016864834.1 |
Set Membership: |
HapMap |