Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAAGAATCTGTCATTCAAATT[A/C]TCCTGGCTTCTTTAGTGGTGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609361 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPE1-MOB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPE1-MOB4 - HSPE1-MOB4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202485.1 | Intron | NP_001189414.1 |
MOB4 - MOB family member 4, phocein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100819.2 | Intron | NP_001094289.1 | ||||
NM_001204094.1 | Intron | NP_001191023.1 | ||||
NM_015387.4 | Intron | NP_056202.2 | ||||
NM_199482.3 | Intron | NP_955776.1 |