Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATTTTCTACTTTTCTTCCATGA[G/T]AACAACATATTAATGTTAACACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611655 | ||||||||||||||||||||
Literature Links: |
PGAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGAP1 - post-GPI attachment to proteins 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321099.1 | 8788 | UTR 3 | NP_001308028.1 | |||
NM_001321100.1 | 8788 | UTR 3 | NP_001308029.1 | |||
NM_024989.3 | 8788 | UTR 3 | NP_079265.2 | |||
XM_017004992.1 | 8788 | UTR 3 | XP_016860481.1 | |||
XM_017004993.1 | 8788 | UTR 3 | XP_016860482.1 | |||
XM_017004994.1 | 8788 | UTR 3 | XP_016860483.1 |