Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGTGTGTACTTGAGTATCTCA[A/G]CAACTGAGGACCGGAAGCGAGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600239 | ||||||||||||||||||||
Literature Links: |
GPR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR1 - G protein-coupled receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098199.1 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | NP_001091669.1 | |
NM_001261452.1 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | NP_001248381.1 | |
NM_001261453.1 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | NP_001248382.1 | |
NM_001261454.1 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | NP_001248383.1 | |
NM_001261455.1 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | NP_001248384.1 | |
NM_005279.3 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | NP_005270.2 | |
XM_005246471.2 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | XP_005246528.1 | |
XM_017003832.1 | 1183 | Missense Mutation | GCT,GTT | A,V 320 | XP_016859321.1 |