Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACATATGGAAGGTTAAAAATC[C/T]ACCCCACTTCTAGTGCTTAGTACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL4A3 - collagen type IV alpha 3 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000091.4 | Intron | NP_000082.2 | ||||
XM_005246277.3 | Intron | XP_005246334.1 | ||||
XM_005246280.3 | Intron | XP_005246337.1 | ||||
XM_006712245.3 | Intron | XP_006712308.1 | ||||
XM_011510555.1 | Intron | XP_011508857.1 | ||||
XM_011510556.2 | Intron | XP_011508858.1 | ||||
XM_017003295.1 | Intron | XP_016858784.1 |
Set Membership: |
HapMap |