Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGGTACACATTTCCATCATGTG[A/C]CCCATAACGTTTGAGATGGGTGTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605153 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAMP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAMP1 - receptor activity modifying protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308353.1 | Intron | NP_001295282.1 | ||||
NM_005855.3 | Intron | NP_005846.1 | ||||
XM_017003152.1 | Intron | XP_016858641.1 | ||||
XM_017003153.1 | Intron | XP_016858642.1 | ||||
XM_017003154.1 | Intron | XP_016858643.1 | ||||
XM_017003155.1 | Intron | XP_016858644.1 | ||||
XM_017003156.1 | Intron | XP_016858645.1 |
Set Membership: |
HapMap |