Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTAAAAGAAAAAGGGTATTTTGT[C/T]TGTCACTTTAAAGCTGCTTTTTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613897 MIM: 602835 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCF PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FANCF - Fanconi anemia complementation group F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022725.3 | 1475 | UTR 3 | NP_073562.1 |
GAS2 - growth arrest specific 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |