Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCAGACTCTTCAATAGGACAA[C/T]ATCATAAATAAGGAACTATTCTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615933 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTBD10 - BTB domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297741.1 | Intron | NP_001284670.1 | ||||
NM_001297742.1 | Intron | NP_001284671.1 | ||||
NM_032320.6 | Intron | NP_115696.2 | ||||
XM_017018405.1 | Intron | XP_016873894.1 | ||||
XM_017018406.1 | Intron | XP_016873895.1 | ||||
XM_017018407.1 | Intron | XP_016873896.1 | ||||
XM_017018408.1 | Intron | XP_016873897.1 | ||||
XM_017018409.1 | Intron | XP_016873898.1 | ||||
XM_017018410.1 | Intron | XP_016873899.1 | ||||
XM_017018411.1 | Intron | XP_016873900.1 |