Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTCTACACGCACTGTTCTAAG[C/G]ATTCAAGATACAACAAAGAAAAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604353 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHORDC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHORDC1 - cysteine and histidine rich domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144073.1 | Intron | NP_001137545.1 | ||||
NM_012124.2 | Intron | NP_036256.2 | ||||
XM_017017540.1 | Intron | XP_016873029.1 | ||||
XM_017017541.1 | Intron | XP_016873030.1 |
Set Membership: |
HapMap |