Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTATTAACAGGTTATATTTTT[C/T]CATTTGTTTAGGATGTCTTTAGTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609766 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWC15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CWC15 - CWC15 spliceosome-associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KDM4D - lysine demethylase 4D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018039.2 | Intron | NP_060509.2 |
Set Membership: |
HapMap |