Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGCCCCCCTTCACCCAGCCTTC[A/G]CCACTGTTAACATTTACATACCTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614657 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMOTL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMOTL1 - angiomotin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301007.1 | Intron | NP_001287936.1 | ||||
NM_130847.2 | Intron | NP_570899.1 | ||||
XM_005273798.4 | Intron | XP_005273855.1 | ||||
XM_005273801.4 | Intron | XP_005273858.1 | ||||
XM_006718772.3 | Intron | XP_006718835.1 | ||||
XM_011542626.2 | Intron | XP_011540928.1 | ||||
XM_011542628.2 | Intron | XP_011540930.1 |
LOC105369438 - uncharacterized LOC105369438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |