Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGACAATGACTTCATGGTATA[C/T]ATCAATTCCTATAAAGAAAAAATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602395 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPAM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPAM - glycerol-3-phosphate acyltransferase, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244949.1 | 4322 | UTR 3 | NP_001231878.1 | |||
NM_020918.5 | 4322 | UTR 3 | NP_065969.3 | |||
XM_005269998.1 | 4322 | UTR 3 | XP_005270055.1 |
Set Membership: |
HapMap |