Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTGTGTTTGCACCTGCGATTAG[C/T]GAGTAGGTAGGCACTTAGGTCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606283 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SORCS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SORCS1 - sortilin related VPS10 domain containing receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013031.2 | 6839 | UTR 3 | NP_001013049.1 | |||
NM_001206569.1 | 6839 | Intron | NP_001193498.1 | |||
NM_001206570.1 | 6839 | UTR 3 | NP_001193499.1 | |||
NM_001206571.1 | 6839 | Intron | NP_001193500.1 | |||
NM_001206572.1 | 6839 | UTR 3 | NP_001193501.1 | |||
NM_052918.4 | 6839 | UTR 3 | NP_443150.3 | |||
XM_011539199.2 | 6839 | Intron | XP_011537501.1 | |||
XM_011539201.2 | 6839 | Intron | XP_011537503.1 | |||
XM_017015614.1 | 6839 | Intron | XP_016871103.1 | |||
XM_017015615.1 | 6839 | Intron | XP_016871104.1 | |||
XM_017015616.1 | 6839 | Intron | XP_016871105.1 | |||
XM_017015617.1 | 6839 | Intron | XP_016871106.1 | |||
XM_017015618.1 | 6839 | Intron | XP_016871107.1 |
Set Membership: |
HapMap |