Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAGGGCTGGCAGATGCCACCA[G/T]GGGTGTGAACGTGAGGGATGCCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612314 | ||||||||||||||||||||
Literature Links: |
GSTO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTO2 - glutathione S-transferase omega 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPRIP - inositol 1,4,5-trisphosphate receptor interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272012.1 | 1847 | UTR 3 | NP_001258941.1 | |||
NM_001272013.1 | 1847 | UTR 3 | NP_001258942.1 | |||
NM_033397.3 | 1847 | UTR 3 | NP_203755.1 | |||
XM_005270257.2 | 1847 | UTR 3 | XP_005270314.1 |
LOC101927472 - uncharacterized LOC101927472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |