Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGAATGGAAGGAATCTAGGCA[A/G]ACTCCTAAATTTGGGGTCTGATCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605482 MIM: 612314 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTO1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GSTO1 - glutathione S-transferase omega 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191002.1 | Intron | NP_001177931.1 | ||||
NM_001191003.1 | Intron | NP_001177932.1 | ||||
NM_004832.2 | Intron | NP_004823.1 |
GSTO2 - glutathione S-transferase omega 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4482 - microRNA 4482 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |