Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATTAACTAGAAGGAGAGTTCC[A/T]TGATGAAATATAGTCAGTAAGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607698 | ||||||||||||||||||||
Literature Links: |
LCOR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LCOR - ligand dependent nuclear receptor corepressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170765.1 | Intron | NP_001164236.1 | ||||
NM_001170766.1 | Intron | NP_001164237.1 | ||||
NM_032440.3 | Intron | NP_115816.2 | ||||
XM_006718033.3 | Intron | XP_006718096.1 | ||||
XM_011540277.2 | Intron | XP_011538579.1 | ||||
XM_017016784.1 | Intron | XP_016872273.1 | ||||
XM_017016785.1 | Intron | XP_016872274.1 | ||||
XM_017016786.1 | Intron | XP_016872275.1 | ||||
XM_017016787.1 | Intron | XP_016872276.1 | ||||
XM_017016788.1 | Intron | XP_016872277.1 |