Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGAGATTAGCTTCAGCCACAA[C/G]AAGTTCATTGGTGTCTTTAATATAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136131 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FMO4 - flavin containing monooxygenase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002022.2 | Intron | NP_002013.1 | ||||
XM_005245045.3 | Intron | XP_005245102.1 | ||||
XM_005245046.3 | Intron | XP_005245103.1 | ||||
XM_005245048.3 | Intron | XP_005245105.1 | ||||
XM_006711243.3 | Intron | XP_006711306.1 | ||||
XM_011509347.2 | Intron | XP_011507649.1 | ||||
XM_011509348.2 | Intron | XP_011507650.1 | ||||
XM_011509349.2 | Intron | XP_011507651.1 |
Set Membership: |
HapMap |