Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCATATCAGCAGAGACCATATA[A/G]AGGGCTCTCACTGCAGGCTGACTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMO2 - flavin containing monooxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301347.1 | Intron | NP_001288276.1 | ||||
NM_001460.4 | Intron | NP_001451.2 |