Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGAGAAGGGATGCTGAGTTACA[A/G]TCATTGGTGAAGTTATTGAGATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609908 MIM: 605667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC4 - apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203454.2 | 2260 | UTR 3 | NP_982279.1 |
RGL1 - ral guanine nucleotide dissociation stimulator like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297669.1 | 2260 | Intron | NP_001284598.1 | |||
NM_001297670.1 | 2260 | Intron | NP_001284599.1 | |||
NM_001297671.1 | 2260 | Intron | NP_001284600.1 | |||
NM_001297672.1 | 2260 | Intron | NP_001284601.1 | |||
NM_015149.4 | 2260 | Intron | NP_055964.3 | |||
XM_011509339.2 | 2260 | Intron | XP_011507641.1 | |||
XM_011509341.1 | 2260 | Intron | XP_011507643.1 | |||
XM_011509342.2 | 2260 | Intron | XP_011507644.1 | |||
XM_017000756.1 | 2260 | Intron | XP_016856245.1 |