Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCACCACACAGAGCTGGCAGCGA[C/T]TCCCATGAGTCCCCACCTCCTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605667 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COLGALT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COLGALT2 - collagen beta(1-O)galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303420.1 | Intron | NP_001290349.1 | ||||
NM_001303421.1 | Intron | NP_001290350.1 | ||||
NM_015101.3 | Intron | NP_055916.1 | ||||
XM_011509332.2 | Intron | XP_011507634.1 | ||||
XM_011509334.2 | Intron | XP_011507636.1 | ||||
XM_011509335.2 | Intron | XP_011507637.1 | ||||
XM_011509336.2 | Intron | XP_011507638.1 | ||||
XM_011509337.2 | Intron | XP_011507639.1 | ||||
XM_011509338.2 | Intron | XP_011507640.1 | ||||
XM_017000750.1 | Intron | XP_016856239.1 | ||||
XM_017000751.1 | Intron | XP_016856240.1 |
RGL1 - ral guanine nucleotide dissociation stimulator like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |