Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCGGGTGTCTCCTCTGTCAAG[A/C]AACACTGCGAGTCTGTGAGTCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604283 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRG4 - proteoglycan 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127708.2 | Intron | NP_001121180.2 | ||||
NM_001127709.2 | Intron | NP_001121181.2 | ||||
NM_001127710.2 | Intron | NP_001121182.2 | ||||
NM_001303232.1 | Intron | NP_001290161.1 | ||||
NM_005807.4 | Intron | NP_005798.3 | ||||
XM_017000002.1 | Intron | XP_016855491.1 | ||||
XM_017000003.1 | Intron | XP_016855492.1 |