Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCAAAAACTCTCATTCCATAAGG[G/T]GACAAATTTTACTTCTGAGAGGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611673 | ||||||||||||||||||||
Literature Links: |
TRMT1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRMT1L - tRNA methyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202423.1 | 4737 | UTR 3 | NP_001189352.1 | |||
NM_030934.4 | 4737 | UTR 3 | NP_112196.3 | |||
XM_011510028.2 | 4737 | UTR 3 | XP_011508330.1 | |||
XM_017002419.1 | 4737 | Intron | XP_016857908.1 |