Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTCTTGTAAGCGTTTTATATGC[A/G]TTATTTATTTTAATTCTGAGAATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613775 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAMSAP2 - calmodulin regulated spectrin associated protein family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297707.1 | Intron | NP_001284636.1 | ||||
NM_001297708.1 | Intron | NP_001284637.1 | ||||
NM_203459.2 | Intron | NP_982284.1 | ||||
XM_005245041.3 | Intron | XP_005245098.1 | ||||
XM_017000799.1 | Intron | XP_016856288.1 |