Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTGGGAATTTAATTATAAATA[C/T]AAGTTCCTATATGGGTAACTAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611279 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF14 - kinesin family member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305792.1 | Intron | NP_001292721.1 | ||||
NM_014875.2 | Intron | NP_055690.1 | ||||
XM_011510231.2 | Intron | XP_011508533.1 | ||||
XM_011510232.2 | Intron | XP_011508534.1 | ||||
XM_011510233.2 | Intron | XP_011508535.1 | ||||
XM_011510235.2 | Intron | XP_011508537.1 | ||||
XM_011510236.1 | Intron | XP_011508538.1 | ||||
XM_017003005.1 | Intron | XP_016858494.1 | ||||
XM_017003006.1 | Intron | XP_016858495.1 | ||||
XM_017003007.1 | Intron | XP_016858496.1 |