Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611864 MIM: 609080 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMC10 - armadillo repeat containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161009.2 | Intron | NP_001154481.1 | ||||
NM_001161010.2 | Intron | NP_001154482.1 | ||||
NM_001161011.2 | Intron | NP_001154483.1 | ||||
NM_001161012.2 | Intron | NP_001154484.1 | ||||
NM_001161013.2 | Intron | NP_001154485.1 | ||||
NM_031905.4 | Intron | NP_114111.2 | ||||
XM_005250628.2 | Intron | XP_005250685.1 | ||||
XM_011516601.2 | Intron | XP_011514903.1 | ||||
XM_011516602.2 | Intron | XP_011514904.1 | ||||
XM_017012684.1 | Intron | XP_016868173.1 | ||||
XM_017012685.1 | Intron | XP_016868174.1 | ||||
XM_017012686.1 | Intron | XP_016868175.1 | ||||
XM_017012687.1 | Intron | XP_016868176.1 | ||||
XM_017012688.1 | Intron | XP_016868177.1 | ||||
XM_017012689.1 | Intron | XP_016868178.1 |
FBXL13 - F-box and leucine rich repeat protein 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |