Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTGAGCTAGAGAGGAACAGCAG[A/G]TAGGTGGTATTTGAAGCCATAAGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611864 MIM: 612334 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMC10 - armadillo repeat containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAPEPLD - N-acyl phosphatidylethanolamine phospholipase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122838.1 | Intron | NP_001116310.1 | ||||
NM_198990.4 | Intron | NP_945341.3 | ||||
XM_005250214.4 | Intron | XP_005250271.2 | ||||
XM_005250215.2 | Intron | XP_005250272.1 | ||||
XM_005250218.2 | Intron | XP_005250275.1 | ||||
XM_011515933.2 | Intron | XP_011514235.1 | ||||
XM_011515934.2 | Intron | XP_011514236.1 | ||||
XM_017011854.1 | Intron | XP_016867343.1 | ||||
XM_017011855.1 | Intron | XP_016867344.1 | ||||
XM_017011856.1 | Intron | XP_016867345.1 | ||||
XM_017011857.1 | Intron | XP_016867346.1 |
Set Membership: |
HapMap |