Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATCTGAAAAAATGAAATAAAAA[A/T]TAGACAGATATTTATATAACACTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF800 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF800 - zinc finger protein 800 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176814.4 | 5952 | Intron | NP_789784.2 | |||
XM_005250181.3 | 5952 | Intron | XP_005250238.1 | |||
XM_006715867.3 | 5952 | UTR 3 | XP_006715930.1 | |||
XM_006715868.3 | 5952 | UTR 3 | XP_006715931.1 | |||
XM_006715869.3 | 5952 | UTR 3 | XP_006715932.1 | |||
XM_011515859.2 | 5952 | UTR 3 | XP_011514161.1 |