Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAATAAAACCAGACATTATAAA[C/T]GGGCCTCAGCAATAGTGTTTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSMEM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SSMEM1 - serine rich single-pass membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145268.3 | Intron | NP_660311.1 | ||||
XM_005250141.3 | Intron | XP_005250198.1 | ||||
XM_011515794.2 | Intron | XP_011514096.1 | ||||
XM_011515795.2 | Intron | XP_011514097.1 |
TMEM209 - transmembrane protein 209 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |