Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGAGAGAAAAAAGTAAGGAAA[A/C]CCATATTAAGAACCCAAAACAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604355 MIM: 601029 | ||||||||||||||||||||
Literature Links: |
COPG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPG2 - coatomer protein complex subunit gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290033.1 | Intron | NP_001276962.1 | ||||
NM_012133.5 | Intron | NP_036265.3 |
MEST - mesoderm specific transcript | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253900.1 | Intron | NP_001240829.1 | ||||
NM_001253901.1 | Intron | NP_001240830.1 | ||||
NM_001253902.1 | Intron | NP_001240831.1 | ||||
NM_002402.3 | Intron | NP_002393.2 | ||||
NM_177524.2 | Intron | NP_803490.1 | ||||
NM_177525.2 | Intron | NP_803491.1 | ||||
XM_011516222.1 | Intron | XP_011514524.1 | ||||
XM_017012218.1 | Intron | XP_016867707.1 |