Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616520 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCYL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AHCYL2 - adenosylhomocysteinase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130720.2 | Intron | NP_001124192.1 | ||||
NM_001130722.2 | Intron | NP_001124194.2 | ||||
NM_001130723.2 | Intron | NP_001124195.1 | ||||
NM_015328.3 | Intron | NP_056143.1 | ||||
XM_005250237.1 | Intron | XP_005250294.1 | ||||
XM_011515987.2 | Intron | XP_011514289.1 | ||||
XM_011515988.2 | Intron | XP_011514290.1 | ||||
XM_017011904.1 | Intron | XP_016867393.1 | ||||
XM_017011905.1 | Intron | XP_016867394.1 | ||||
XM_017011906.1 | Intron | XP_016867395.1 | ||||
XM_017011907.1 | Intron | XP_016867396.1 |
Set Membership: |
HapMap |