Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCTAAGTTTTGCATATCATTA[A/G]TTCTACTTGCTCCACTTTTGATAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600160 MIM: 600431 MIM: 613149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN2A - cyclin dependent kinase inhibitor 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDKN2B - cyclin dependent kinase inhibitor 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004936.3 | Intron | NP_004927.2 | ||||
NM_078487.2 | Intron | NP_511042.1 |
CDKN2B-AS1 - CDKN2B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |