Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTTTTGCAATTTCTTATGGGTC[A/C]AAATTATCCCTACATAAAAAGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615941 | ||||||||||||||||||||
Literature Links: |
HACD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HACD4 - 3-hydroxyacyl-CoA dehydratase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010915.4 | Intron | NP_001010915.2 | ||||
NM_001321883.1 | Intron | NP_001308812.1 | ||||
NM_001321903.1 | Intron | NP_001308832.1 | ||||
XM_011517876.2 | Intron | XP_011516178.1 | ||||
XM_017014713.1 | Intron | XP_016870202.1 | ||||
XM_017014714.1 | Intron | XP_016870203.1 |