Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAAAGCATAAATGTTATAAAT[A/C]TTTTTTTAAATTATGGAAAGAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND4C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DENND4C - DENN domain containing 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017925.6 | Intron | NP_060395.5 | ||||
XM_006716804.3 | Intron | XP_006716867.1 | ||||
XM_006716805.3 | Intron | XP_006716868.1 | ||||
XM_006716807.3 | Intron | XP_006716870.1 | ||||
XM_011517959.2 | Intron | XP_011516261.1 | ||||
XM_017014917.1 | Intron | XP_016870406.1 | ||||
XM_017014918.1 | Intron | XP_016870407.1 | ||||
XM_017014919.1 | Intron | XP_016870408.1 | ||||
XM_017014920.1 | Intron | XP_016870409.1 |