Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATATAGGAAATTCCAGAGCCAGA[G/T]AGTATAAAATATTTATTGAAAACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613433 MIM: 612194 MIM: 616292 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HAUS6 - HAUS augmin like complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270890.1 | 5207 | Intron | NP_001257819.1 | |||
NM_017645.4 | 5207 | Intron | NP_060115.3 | |||
XM_011517935.2 | 5207 | UTR 3 | XP_011516237.1 | |||
XM_017014830.1 | 5207 | Intron | XP_016870319.1 |
RRAGA - Ras related GTP binding A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAXO1 - stabilizer of axonemal microtubules 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |