Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATGTTAATGTCACATTCTCCT[A/G]AAGTGTGATATGATAGAAAGACCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601899 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STAM - signal transducing adaptor molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324282.1 | Intron | NP_001311211.1 | ||||
NM_001324283.1 | Intron | NP_001311212.1 | ||||
NM_001324284.1 | Intron | NP_001311213.1 | ||||
NM_001324285.1 | Intron | NP_001311214.1 | ||||
NM_001324286.1 | Intron | NP_001311215.1 | ||||
NM_001324287.1 | Intron | NP_001311216.1 | ||||
NM_001324288.1 | Intron | NP_001311217.1 | ||||
NM_001324289.1 | Intron | NP_001311218.1 | ||||
NM_003473.3 | Intron | NP_003464.1 | ||||
XM_011519695.2 | Intron | XP_011517997.1 | ||||
XM_017016685.1 | Intron | XP_016872174.1 |
STAM-AS1 - STAM antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |