Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGTATCCCCTGCGGGAAAGACCA[C/T]GCTGGCTGCTGACCAAACTCCACTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611380 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIP2C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIP2C - disco interacting protein 2 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014974.2 | Intron | NP_055789.1 | ||||
XM_005252426.3 | Intron | XP_005252483.1 | ||||
XM_005252427.3 | Intron | XP_005252484.1 | ||||
XM_005252428.4 | Intron | XP_005252485.1 | ||||
XM_005252430.2 | Intron | XP_005252487.2 | ||||
XM_011519428.2 | Intron | XP_011517730.1 | ||||
XM_011519429.2 | Intron | XP_011517731.1 | ||||
XM_011519430.2 | Intron | XP_011517732.1 | ||||
XM_011519431.2 | Intron | XP_011517733.1 | ||||
XM_011519432.1 | Intron | XP_011517734.1 | ||||
XM_011519433.2 | Intron | XP_011517735.1 | ||||
XM_017015977.1 | Intron | XP_016871466.1 |
PRR26 - proline rich 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |