Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCGTCCTATGGCACCTATGTT[A/G]TAAACGTTTTGAAGAATAGCCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616257 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FKBP9 - FK506 binding protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284341.1 | Intron | NP_001271270.1 | ||||
NM_001284343.1 | Intron | NP_001271272.1 | ||||
NM_007270.4 | Intron | NP_009201.2 | ||||
XM_011515115.2 | Intron | XP_011513417.1 |