Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGATCCAGAGAGTTTGCAAGA[C/G]AGAGGCGAAGGGAAGGTCGCAACAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606860 | |||||||||||||||||||||||
Literature Links: |
SERPING1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SERPING1 - serpin family G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000062.2 | 156 | Missense Mutation | GAC,GAG | D,E 39 | NP_000053.2 | |
NM_001032295.1 | 156 | Missense Mutation | GAC,GAG | D,E 39 | NP_001027466.1 |
Set Membership: |
HapMap |