Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCAACAGTACCAAAGCCCTCAC[A/G]TGTGGCCGCCCATTCTCGCACCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608757 MIM: 610175 MIM: 609725 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLP1 - cleavage and polyadenylation factor I subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR130A - microRNA 130a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YPEL4 - yippee like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145008.2 | 2018 | Intron | NP_659445.1 | |||
XM_017017318.1 | 2018 | UTR 5 | XP_016872807.1 |