Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTACATGACTTCTAAACTTGCTT[C/T]TTAATATTCCAGCAAGAAAAAATGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPXN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LPXN - leupaxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143995.2 | Intron | NP_001137467.1 | ||||
NM_001307951.1 | Intron | NP_001294880.1 | ||||
NM_004811.2 | Intron | NP_004802.1 | ||||
XM_006718750.1 | Intron | XP_006718813.1 | ||||
XM_011545388.2 | Intron | XP_011543690.1 | ||||
XM_011545390.2 | Intron | XP_011543692.1 | ||||
XM_011545391.2 | Intron | XP_011543693.1 | ||||
XM_011545393.2 | Intron | XP_011543695.1 | ||||
XM_011545394.2 | Intron | XP_011543696.1 | ||||
XM_017018579.1 | Intron | XP_016874068.1 |
Set Membership: |
HapMap |