Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCCCATTCTTATATATATTTGT[A/G]TATAGAAAAGGCTTTGGAAGGGTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602647 MIM: 603189 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NXF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NXF1 - nuclear RNA export factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX5 - syntaxin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244666.1 | Intron | NP_001231595.1 | ||||
NM_003164.4 | Intron | NP_003155.2 | ||||
XM_011545223.2 | Intron | XP_011543525.1 | ||||
XM_011545224.2 | Intron | XP_011543526.1 |
Set Membership: |
HapMap |