Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGAAATGCCATCTCCCTTACT[C/T]TAATGTTGCTATATTAATATGTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605693 | ||||||||||||||||||||
Literature Links: |
RAB30 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB30 - RAB30, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286059.1 | 5059 | UTR 3 | NP_001272988.1 | |||
NM_001286060.1 | 5059 | UTR 3 | NP_001272989.1 | |||
NM_001286061.1 | 5059 | UTR 3 | NP_001272990.1 | |||
NM_014488.4 | 5059 | UTR 3 | NP_055303.2 |